Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frtz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272950.1 | NP_001259879.1 | 4 - 5 | 2473 | 97 | |
NM_134770.2 | NP_608614.1 | 4 - 5 | 2473 | 97 | ||
GenBank mRNA | BT004832.1 | - | 4 - 5 | 2485 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |