Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mio | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134812.3 | NP_608656.1 | 6 - 7 | 2647 | 74 | |
GenBank mRNA | AY061485.1 | - | 6 - 7 | 2574 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |