Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGlut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258919.1 | NP_001245848.1 | 3 - 4 | 849 | 93 | |
NM_134837.3 | NP_608681.2 | 3 - 4 | 690 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |