Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnpo-SR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273032.1 | NP_001259961.1 | 2 - 3 | 1315 | 82 | |
NM_134864.4 | NP_608708.2 | 2 - 3 | 1315 | 82 | ||
GenBank mRNA | AY095186.1 | - | 2 - 3 | 1270 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |