Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134901.2 | NP_608745.1 | 1 - 2 | 176 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |