Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079909.3 | NP_524648.3 | 3 - 4 | 1687 | 72 | |
NM_134924.2 | NP_608768.2 | 3 - 4 | 2439 | 72 | ||
NM_205900.2 | NP_995622.1 | 3 - 4 | 1804 | 72 | ||
GenBank mRNA | BT133432.1 | - | 3 - 4 | 1804 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |