Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgant2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134929.4 | NP_608773.2 | 6 - 7 | 1746 | 56 | |
NM_205903.3 | NP_995625.2 | 6 - 7 | 1782 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |