Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: teq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169922.3 | NP_001163393.2 | 9 - 10 | 2928 | 116 | |
NM_140031.3 | NP_648288.2 | 9 - 10 | 2928 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |