Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGRP-LF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140042.3 | NP_648299.3 | 2 - 3 | 359 | 93 | |
GenBank mRNA | AY084190.1 | - | 2 - 3 | 357 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |