Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTF-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104090.2 | NP_001097560.1 | 3 - 4 | 1273 | 69 | |
NM_001104091.2 | NP_001097561.1 | 2 - 3 | 1125 | 69 | ||
NM_001300073.1 | NP_001287002.1 | 3 - 4 | 1273 | 69 | ||
NM_001300074.1 | NP_001287003.1 | 3 - 4 | 776 | 69 | ||
NM_140054.3 | NP_648311.2 | 2 - 3 | 628 | 69 | ||
GenBank mRNA | BT029122.1 | - | 2 - 3 | 1093 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |