Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: path | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274705.1 | NP_001261634.1 | 4 - 5 | 665 | 66 | |
NM_140070.3 | NP_648327.1 | 3 - 4 | 315 | 66 | ||
GenBank mRNA | AY070669.1 | - | 3 - 4 | 317 | 66 | |
AY070688.1 | - | 3 - 4 | 317 | 66 | ||
BT088839.1 | - | 3 - 4 | 289 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |