Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or67d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140133.2 | NP_648390.2 | 1 - 2 | 333 | 92 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Drosophila melanogaster |