Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169942.2 | NP_001163413.1 | 10 - 11 | 3865 | 65 | |
NM_001382194.1 | NP_001368945.1 | 10 - 11 | 3432 | 65 | ||
NM_140155.4 | NP_648412.2 | 10 - 11 | 4012 | 65 | ||
NM_168422.3 | NP_729629.2 | 9 - 10 | 3427 | 65 | ||
GenBank mRNA | BT003272.1 | - | 10 - 11 | 3865 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |