Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140164.4 | NP_648421.1 | 2 - 3 | 775 | 67 | |
NM_168425.3 | NP_729639.1 | 2 - 3 | 1245 | 67 | ||
NM_168426.3 | NP_729640.1 | 3 - 4 | 954 | 67 | ||
GenBank mRNA | AJ863561.1 | - | 3 - 4 | 482 | 67 | |
AJ863564.1 | - | 2 - 3 | 404 | 67 | ||
AY118969.1 | - | 3 - 4 | 481 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |