Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elo68beta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104110.4 | NP_001097580.1 | 3 - 4 | 531 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |