Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FoxK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274772.2 | NP_001261701.1 | 8 - 9 | 2139 | 151 | |
NM_001274773.2 | NP_001261702.1 | 8 - 9 | 2136 | 151 | ||
NM_140183.3 | NP_648440.2 | 8 - 9 | 2139 | 151 | ||
NM_168443.3 | NP_729672.1 | 8 - 9 | 2121 | 151 | ||
NM_168444.2 | NP_729673.2 | 8 - 9 | 2220 | 151 | ||
NM_168445.3 | NP_729674.2 | 8 - 9 | 2017 | 151 | ||
NM_168446.2 | NP_729675.2 | 8 - 9 | 2095 | 151 | ||
GenBank mRNA | AY787838.1 | - | 7 - 8 | 1947 | 151 | |
BT023507.1 | - | 8 - 9 | 2075 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |