Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140195.4 | NP_648452.1 | 1 - 2 | 427 | 61 | |
GenBank mRNA | AY069489.1 | - | 1 - 2 | 372 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |