Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir68a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300102.1 | NP_001287031.1 | 3 - 4 | 1211 | 68 | |
NM_140198.3 | NP_648455.2 | 3 - 4 | 1211 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |