Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140329.3 | NP_648586.2 | 2 - 3 | 2017 | 76 | |
GenBank mRNA | AY069097.1 | - | 2 - 3 | 1995 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |