Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273193.1 | NP_001260122.1 | 3 - 4 | 727 | 82 | |
NM_164683.3 | NP_723157.1 | 1 - 2 | 555 | 82 | ||
NM_164684.3 | NP_723158.1 | 1 - 2 | 555 | 82 | ||
GenBank mRNA | AY061071.1 | - | 1 - 2 | 541 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |