Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273193.1 | NP_001260122.1 | 4 - 5 | 810 | 66 | |
NM_164683.3 | NP_723157.1 | 2 - 3 | 638 | 66 | ||
NM_164684.3 | NP_723158.1 | 2 - 3 | 638 | 66 | ||
GenBank mRNA | AY061071.1 | - | 2 - 3 | 624 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |