Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273248.1 | NP_001260177.1 | 2 - 3 | 613 | 79 | |
NM_001273249.1 | NP_001260178.1 | 2 - 3 | 613 | 79 | ||
NM_001273253.1 | NP_001260182.1 | 2 - 3 | 420 | 79 | ||
NM_164743.3 | NP_723263.2 | 2 - 3 | 420 | 79 | ||
NM_164744.2 | NP_723264.1 | 2 - 3 | 420 | 79 | ||
GenBank mRNA | BT128745.1 | - | 2 - 3 | 417 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |