Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135291.3 | NP_609135.1 | 4 - 5 | 797 | 138 | |
NM_164765.4 | NP_723303.1 | 4 - 5 | 588 | 138 | ||
GenBank mRNA | BT133486.1 | - | 4 - 5 | 585 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |