Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144336.3 | NP_001137808.1 | 3 - 4 | 915 | 76 | |
NM_001298802.1 | NP_001285731.1 | 3 - 4 | 797 | 76 | ||
NM_001298803.1 | NP_001285732.1 | 3 - 4 | 653 | 76 | ||
NM_135324.3 | NP_609168.2 | 2 - 3 | 532 | 76 | ||
NM_164777.3 | NP_723328.2 | 3 - 4 | 646 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |