Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259007.2 | NP_001245936.1 | 2 - 3 | 700 | 73 | |
NM_001259008.2 | NP_001245937.1 | 2 - 3 | 880 | 73 | ||
NM_001273292.1 | NP_001260221.1 | 2 - 3 | 700 | 73 | ||
NM_135333.3 | NP_609177.1 | 2 - 3 | 1167 | 73 | ||
NM_164779.3 | NP_723334.1 | 2 - 3 | 700 | 73 | ||
NM_164780.3 | NP_723335.1 | 2 - 3 | 1147 | 73 | ||
NM_164781.3 | NP_723336.1 | 2 - 3 | 896 | 73 | ||
GenBank mRNA | AF364305.1 | - | 2 - 3 | 1149 | 73 | |
AY061460.1 | - | 2 - 3 | 674 | 73 | ||
BT120179.1 | - | 2 - 3 | 673 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |