Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273292.1 | NP_001260221.1 | 4 - 5 | 1371 | 68 | |
NM_135333.3 | NP_609177.1 | 4 - 5 | 1862 | 68 | ||
NM_164779.3 | NP_723334.1 | 4 - 5 | 1395 | 68 | ||
NM_164780.3 | NP_723335.1 | 4 - 5 | 1842 | 68 | ||
NM_164781.3 | NP_723336.1 | 4 - 5 | 1591 | 68 | ||
GenBank mRNA | AF364305.1 | - | 4 - 5 | 1844 | 68 | |
AY061460.1 | - | 4 - 5 | 1369 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |