Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piezo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042881.5 | NP_001036346.3 | 27 - 28 | 7749 | 79 | |
NM_001169436.3 | NP_001162907.1 | 27 - 28 | 7794 | 79 | ||
NM_001201789.2 | NP_001188718.1 | 26 - 27 | 7446 | 79 | ||
NM_001201790.2 | NP_001188719.1 | 25 - 26 | 7389 | 79 | ||
NM_001273297.1 | NP_001260226.1 | 26 - 27 | 7449 | 79 | ||
NM_001273298.1 | NP_001260227.1 | 25 - 26 | 7389 | 79 | ||
NM_001273299.1 | NP_001260228.1 | 24 - 25 | 7335 | 79 | ||
NM_001273300.2 | NP_001260229.2 | 26 - 27 | 8250 | 79 | ||
NM_001316385.1 | NP_001303314.1 | 28 - 29 | 8655 | 79 | ||
NM_135344.6 | NP_609188.4 | 27 - 28 | 7538 | 79 | ||
NM_164795.6 | NP_723354.3 | 27 - 28 | 7863 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |