Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piezo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042881.5 | NP_001036346.3 | 28 - 29 | 8253 | 70 | |
NM_001169436.3 | NP_001162907.1 | 28 - 29 | 8298 | 70 | ||
NM_001201789.2 | NP_001188718.1 | 27 - 28 | 7950 | 70 | ||
NM_001201790.2 | NP_001188719.1 | 26 - 27 | 7893 | 70 | ||
NM_001273297.1 | NP_001260226.1 | 27 - 28 | 7953 | 70 | ||
NM_001273298.1 | NP_001260227.1 | 26 - 27 | 7893 | 70 | ||
NM_001273299.1 | NP_001260228.1 | 25 - 26 | 7839 | 70 | ||
NM_001273300.2 | NP_001260229.2 | 27 - 28 | 8754 | 70 | ||
NM_001316385.1 | NP_001303314.1 | 29 - 30 | 9159 | 70 | ||
NM_135344.6 | NP_609188.4 | 28 - 29 | 8042 | 70 | ||
NM_164795.6 | NP_723354.3 | 28 - 29 | 8367 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |