Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piezo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042881.5 | NP_001036346.3 | 5 - 6 | 1339 | 60 | |
NM_001169436.3 | NP_001162907.1 | 5 - 6 | 1339 | 60 | ||
NM_001201789.2 | NP_001188718.1 | 5 - 6 | 1339 | 60 | ||
NM_001201790.2 | NP_001188719.1 | 5 - 6 | 1339 | 60 | ||
NM_001273297.1 | NP_001260226.1 | 5 - 6 | 1339 | 60 | ||
NM_001273298.1 | NP_001260227.1 | 5 - 6 | 1339 | 60 | ||
NM_001273299.1 | NP_001260228.1 | 5 - 6 | 1339 | 60 | ||
NM_001273300.2 | NP_001260229.2 | 5 - 6 | 1339 | 60 | ||
NM_001316385.1 | NP_001303314.1 | 5 - 6 | 1339 | 60 | ||
NM_135344.6 | NP_609188.4 | 5 - 6 | 1278 | 60 | ||
NM_164795.6 | NP_723354.3 | 5 - 6 | 1339 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |