Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piezo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042881.5 | NP_001036346.3 | 7 - 8 | 1621 | 76 | |
NM_001273297.1 | NP_001260226.1 | 7 - 8 | 1621 | 76 | ||
NM_164795.6 | NP_723354.3 | 7 - 8 | 1621 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |