Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169438.2 | NP_001162909.1 | 8 - 9 | 2538 | 68 | |
NM_001169439.2 | NP_001162910.1 | 9 - 10 | 2125 | 68 | ||
NM_001169440.2 | NP_001162911.1 | 9 - 10 | 2125 | 68 | ||
NM_001201792.2 | NP_001188721.1 | 8 - 9 | 1794 | 68 | ||
NM_078785.4 | NP_523509.2 | 8 - 9 | 1921 | 68 | ||
NM_164801.3 | NP_723365.1 | 8 - 9 | 1921 | 68 | ||
NM_175983.3 | NP_787997.2 | 8 - 9 | 1903 | 68 | ||
NM_205926.3 | NP_995648.1 | 8 - 9 | 1956 | 68 | ||
NM_205927.3 | NP_995649.1 | 8 - 9 | 2538 | 68 | ||
GenBank mRNA | BT050546.1 | - | 7 - 8 | 1670 | 68 | |
BT133154.1 | - | 9 - 10 | 2094 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |