Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316387.1 | NP_001303316.1 | 6 - 7 | 3411 | 94 | |
NM_164818.2 | NP_723391.2 | 6 - 7 | 3411 | 94 | ||
GenBank mRNA | AF179360.1 | - | 5 - 6 | 3277 | 94 | |
AJ249178.1 | - | 5 - 6 | 3040 | 94 | ||
AY089558.1 | - | 6 - 7 | 3395 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |