Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oatp30B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135449.4 | NP_609293.2 | 3 - 4 | 652 | 56 | |
NM_164857.3 | NP_723463.1 | 3 - 4 | 707 | 56 | ||
NM_205946.2 | NP_995668.1 | 2 - 3 | 526 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |