Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oatp30B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135449.4 | NP_609293.2 | 6 - 7 | 2936 | 78 | |
NM_164857.3 | NP_723463.1 | 6 - 7 | 2991 | 78 | ||
NM_205945.2 | NP_995667.1 | 4 - 5 | 2332 | 78 | ||
NM_205946.2 | NP_995668.1 | 5 - 6 | 2810 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |