Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nAChRalpha6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201835.3 | NP_001188764.1 | 11 - 12 | 3179 | 59 | |
NM_001273373.2 | NP_001260302.1 | 11 - 12 | 3179 | 59 | ||
NM_135472.6 | NP_609316.4 | 10 - 11 | 2461 | 59 | ||
NM_164874.3 | NP_723494.2 | 11 - 12 | 3385 | 59 | ||
NM_205951.2 | NP_995673.1 | 10 - 11 | 2461 | 59 | ||
NM_205952.3 | NP_995674.1 | 12 - 13 | 3430 | 59 | ||
NM_205953.3 | NP_995675.1 | 11 - 12 | 2342 | 59 | ||
GenBank mRNA | AF321445.1 | - | 9 - 10 | 1392 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |