Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nAChRalpha6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201835.3 | NP_001188764.1 | 7 - 8 | 2706 | 94 | |
NM_001273373.2 | NP_001260302.1 | 7 - 8 | 2706 | 94 | ||
NM_135472.6 | NP_609316.4 | 6 - 7 | 1988 | 94 | ||
NM_164874.3 | NP_723494.2 | 7 - 8 | 2912 | 94 | ||
NM_205951.2 | NP_995673.1 | 6 - 7 | 1988 | 94 | ||
NM_205952.3 | NP_995674.1 | 8 - 9 | 2957 | 94 | ||
NM_205953.3 | NP_995675.1 | 6 - 7 | 1782 | 94 | ||
GenBank mRNA | AF321445.1 | - | 5 - 6 | 919 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |