Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RluA-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273422.1 | NP_001260351.1 | 6 - 7 | 1430 | 83 | |
NM_164930.2 | NP_723592.1 | 7 - 8 | 1788 | 83 | ||
NM_164931.2 | NP_723593.1 | 7 - 8 | 1922 | 83 | ||
NM_164932.2 | NP_723594.1 | 7 - 8 | 1629 | 83 | ||
GenBank mRNA | BT044578.1 | - | 7 - 8 | 1787 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |