Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ge-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135642.3 | NP_609486.2 | 2 - 3 | 511 | 60 | |
NM_164957.2 | NP_723654.1 | 1 - 2 | 594 | 60 | ||
GenBank mRNA | BT004479.1 | - | 2 - 3 | 491 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |