Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir25a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273120.1 | NP_001260049.1 | 4 - 5 | 1460 | 61 | |
NM_001273121.1 | NP_001260050.1 | 3 - 4 | 952 | 61 | ||
NM_001273122.1 | NP_001260051.1 | 4 - 5 | 1153 | 61 | ||
NM_135019.2 | NP_608863.2 | 5 - 6 | 1018 | 61 | ||
GenBank mRNA | HQ600588.1 | - | 4 - 5 | 980 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |