Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hoe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273130.1 | NP_001260059.1 | 3 - 4 | 817 | 95 | |
NM_001273131.1 | NP_001260060.1 | 3 - 4 | 775 | 95 | ||
NM_001298678.1 | NP_001285607.1 | 2 - 3 | 574 | 95 | ||
NM_135031.4 | NP_608875.1 | 2 - 3 | 559 | 95 | ||
NM_135032.2 | NP_608876.1 | 2 - 3 | 492 | 95 | ||
NM_164603.2 | NP_723024.2 | 3 - 4 | 703 | 95 | ||
NM_175966.2 | NP_787980.1 | 3 - 4 | 787 | 95 | ||
NM_175967.2 | NP_787981.1 | 3 - 4 | 825 | 95 | ||
NM_175968.2 | NP_787982.1 | 3 - 4 | 771 | 95 | ||
GenBank mRNA | AF145605.1 | - | 2 - 3 | 538 | 95 | |
AY050571.1 | - | 3 - 4 | 790 | 95 | ||
AY050572.1 | - | 2 - 3 | 541 | 95 | ||
BT016131.1 | - | 3 - 4 | 790 | 95 | ||
BT044574.1 | - | 2 - 3 | 487 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |