Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jon25Biii | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135037.2 | NP_608881.1 | 1 | 741 | 85 | |
GenBank mRNA | AY075474.1 | - | 1 | 739 | 85 | |
AY113592.1 | - | 1 | 740 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |