Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marcal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135039.2 | NP_608883.1 | 6 - 7 | 2129 | 79 | |
GenBank mRNA | BT011033.1 | - | 6 - 7 | 2125 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |