Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4ac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135072.2 | NP_608916.1 | 3 - 4 | 517 | 112 | |
GenBank mRNA | AY051602.1 | - | 3 - 4 | 517 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |