Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273169.1 | NP_001260098.1 | 2 - 3 | 361 | 57 | |
NM_001273170.1 | NP_001260099.1 | 2 - 3 | 346 | 57 | ||
NM_135086.4 | NP_608930.1 | 1 - 2 | 287 | 57 | ||
GenBank mRNA | AF503572.2 | - | 1 - 2 | 270 | 57 | |
AY060849.1 | - | 1 - 2 | 275 | 57 | ||
AY113391.1 | - | 1 - 2 | 285 | 57 | ||
KX531732.1 | - | 1 - 2 | 146 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |