Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap-D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169416.2 | NP_001162887.1 | 4 - 5 | 2019 | 113 | |
NM_135091.4 | NP_608935.2 | 4 - 5 | 2019 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |