Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144477.2 | NP_001137949.1 | 3 - 4 | 2866 | 66 | |
NM_001144478.4 | NP_001137950.1 | 4 - 5 | 2885 | 66 | ||
NM_001144479.4 | NP_001137951.1 | 4 - 5 | 2975 | 66 | ||
NM_001144480.3 | NP_001137952.1 | 4 - 5 | 2877 | 66 | ||
NM_001144481.2 | NP_001137953.1 | 4 - 5 | 4914 | 66 | ||
NM_001259826.1 | NP_001246755.1 | 6 - 7 | 7624 | 66 | ||
NM_140447.4 | NP_648704.2 | 4 - 5 | 2913 | 66 | ||
NM_140448.3 | NP_648705.2 | 6 - 7 | 7608 | 66 | ||
GenBank mRNA | DQ168462.1 | - | 4 - 5 | 4914 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |