Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274953.1 | NP_001261882.1 | 4 - 5 | 3011 | 91 | |
NM_140518.3 | NP_648775.1 | 6 - 7 | 3277 | 91 | ||
NM_168626.3 | NP_730054.1 | 5 - 6 | 3344 | 91 | ||
GenBank mRNA | BT099682.1 | - | 5 - 6 | 3282 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |