Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGAP71E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144484.3 | NP_001137956.1 | 3 - 4 | 1525 | 63 | |
NM_001202183.2 | NP_001189112.1 | 3 - 4 | 1525 | 63 | ||
NM_001274957.1 | NP_001261886.1 | 3 - 4 | 1525 | 63 | ||
NM_001274958.1 | NP_001261887.1 | 3 - 4 | 1525 | 63 | ||
NM_140527.5 | NP_648784.1 | 3 - 4 | 1525 | 63 | ||
NM_168629.4 | NP_730060.1 | 3 - 4 | 1525 | 63 | ||
NM_168630.4 | NP_730061.1 | 3 - 4 | 1525 | 63 | ||
GenBank mRNA | AY061070.1 | - | 3 - 4 | 1505 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |