Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgant8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140543.4 | NP_648800.1 | 2 - 3 | 1224 | 155 | |
GenBank mRNA | AY070966.1 | - | 2 - 3 | 1221 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |