Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pHCl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038934.4 | NP_001034023.2 | 11 - 12 | 2253 | 97 | |
NM_001259852.1 | NP_001246781.1 | 12 - 13 | 2336 | 97 | ||
NM_001274965.1 | NP_001261894.1 | 11 - 12 | 2315 | 97 | ||
NM_001274967.1 | NP_001261896.1 | 10 - 11 | 2240 | 97 | ||
NM_001274968.1 | NP_001261897.1 | 11 - 12 | 2310 | 97 | ||
NM_001274969.1 | NP_001261898.1 | 12 - 13 | 2331 | 97 | ||
NM_001274971.1 | NP_001261900.1 | 11 - 12 | 2310 | 97 | ||
NM_001370024.1 | NP_001356981.1 | 11 - 12 | 2315 | 97 | ||
NM_001370026.1 | NP_001356909.1 | 10 - 11 | 2240 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |